siRNA Details
| Gene Symbol: Bmp2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770127.1 | XP_008768349.1 | 14 | 2264 | ||
| GenBank mRNA | BC098765.1 | - | 2 | 223 | ||
| BC103651.1 | - | 6 | 1059 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bmp2k| Gene Symbol: Bmp2k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008770127.1 | XP_008768349.1 | 14 | 2264 | ||
| GenBank mRNA | BC098765.1 | - | 2 | 223 | ||
| BC103651.1 | - | 6 | 1059 | |||