siRNA Details
| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017496.1 | NP_001017496.1 | 1 | 49 | ||
| XM_006250710.2 | XP_006250772.1 | 1 | 87 | |||
| GenBank mRNA | BC088260.1 | - | 1 | 49 | 7375992 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cxcl13| Gene Symbol: Cxcl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017496.1 | NP_001017496.1 | 1 | 49 | ||
| XM_006250710.2 | XP_006250772.1 | 1 | 87 | |||
| GenBank mRNA | BC088260.1 | - | 1 | 49 | 7375992 | |