siRNA Details
| Gene Symbol: LOC498339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047928.1 | NP_001041393.1 | 3 | 357 | ||
| GenBank mRNA | AY539941.1 | - | 3 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC498339| Gene Symbol: LOC498339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001047928.1 | NP_001041393.1 | 3 | 357 | ||
| GenBank mRNA | AY539941.1 | - | 3 | 357 | ||