siRNA Details
| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017499.1 | NP_001017499.1 | 1 | 812 | ||
| GenBank mRNA | BC085889.1 | - | 1 | 812 | 7192440 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scfd2| Gene Symbol: Scfd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017499.1 | NP_001017499.1 | 1 | 812 | ||
| GenBank mRNA | BC085889.1 | - | 1 | 812 | 7192440 | |