siRNA Details
| Gene Symbol: Tmem156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025138.1 | NP_001020309.1 | 2 | 227 | ||
| XM_008770160.1 | XP_008768382.1 | 3 | 442 | |||
| GenBank mRNA | BC091217.1 | - | 2 | 227 | 7384360 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem156| Gene Symbol: Tmem156 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025138.1 | NP_001020309.1 | 2 | 227 | ||
| XM_008770160.1 | XP_008768382.1 | 3 | 442 | |||
| GenBank mRNA | BC091217.1 | - | 2 | 227 | 7384360 | |