siRNA Details
Gene Symbol: LOC498368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017500.1 | NP_001017500.1 | 2 | 400 | ||
GenBank mRNA | BC079124.1 | - | 2 | 400 | 7126862 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC498368 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017500.1 | NP_001017500.1 | 2 | 400 | ||
GenBank mRNA | BC079124.1 | - | 2 | 400 | 7126862 |