siRNA Details
| Gene Symbol: Selm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115013.1 | NP_001108485.1 | 4, 5 | 336 | ||
| GenBank mRNA | BG663014.1 | - | 4, 5 | 336 | ||
| BI282694.1 | - | 1, 2 | 357 | |||
| CB607044.1 | - | 4, 5 | 308 | |||
| FQ223498.1 | - | 4, 5 | 291 | |||

Selm
Genomic Map