siRNA Details
| Gene Symbol: Hus1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109092.1 | NP_001102562.1 | 1 | 114 | ||
| XM_006251468.1 | XP_006251530.1 | 1 | 109 | |||
| XM_006251470.2 | XP_006251532.1 | 1 | 105 | |||
| XM_008770346.1 | XP_008768568.1 | 1 | 106 | |||

Hus1
Genomic Map