siRNA Details
Gene Symbol: Grb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109093.1 | NP_001102563.1 | 7 | 845 | ||
XM_006251508.2 | XP_006251570.1 | 5 | 726 | |||
XM_008770347.1 | XP_008768569.1 | 6 | 891 | |||
XM_008770348.1 | XP_008768570.1 | 5 | 726 |
