siRNA Details
| Gene Symbol: Sertad2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024903.1 | NP_001020074.2 | 2 | 832 | ||
| XM_006251526.2 | XP_006251588.1 | 3 | 1095 | |||
| XM_006251527.2 | XP_006251589.1 | 2 | 2217 | |||
| GenBank mRNA | BC098047.1 | - | 2 | 832 | 7457109 | |

Sertad2
Genomic Map