siRNA Details
| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025140.2 | NP_001020311.2 | 28 | 3343 | ||
| XM_008770467.1 | XP_008768689.1 | 29 | 3659 | |||
| GenBank mRNA | DN932843.1 | - | 4 | 346 | ||
| DY314798.1 | - | 10 | 707 | |||

Psme4
Genomic Map