siRNA Details
Gene Symbol: LOC498460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047931.1 | NP_001041396.1 | 1 | 19 | ||
GenBank mRNA | AY383710.1 | - | 1 | 19 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC498460 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047931.1 | NP_001041396.1 | 1 | 19 | ||
GenBank mRNA | AY383710.1 | - | 1 | 19 |