siRNA Details
| Gene Symbol: Gnpnat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134756.1 | NP_001128228.1 | 6 | 595 | ||
| NM_001134757.1 | NP_001128229.1 | 5 | 601 | |||
| XM_006251782.2 | XP_006251844.1 | 6 | 612 | |||
| XM_006251783.2 | XP_006251845.1 | 6 | 660 | |||
| XM_006251784.2 | XP_006251846.1 | 6 | 608 | |||
| GenBank mRNA | BC160861.1 | - | 5 | 601 | 7322586 | |

Gnpnat1
Genomic Map