siRNA Details
| Gene Symbol: Ccnb1ip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025141.1 | NP_001020312.1 | 4 | 549 | ||
| XM_006251901.2 | XP_006251963.1 | 5 | 959 | |||
| XM_006251903.2 | XP_006251965.1 | 5 | 1255 | |||
| XM_006251904.1 | XP_006251966.1 | 3 | 436 | |||
| XM_008770563.1 | XP_008768785.1 | 5 | 1410 | |||
| XM_008770564.1 | XP_008768786.1 | 6 | 1307 | |||
| XM_008770565.1 | XP_008768787.1 | 6 | 1180 | |||
| XM_008770566.1 | XP_008768788.1 | 5 | 1241 | |||
| XM_008770567.1 | XP_008768789.1 | 4 | 1062 | |||
| XM_008770568.1 | XP_008768790.1 | 5 | 1818 | |||
| GenBank mRNA | BC091423.1 | - | 4 | 549 | 7365771 | |

Ccnb1ip1
Genomic Map