siRNA Details
| Gene Symbol: Fam167a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109102.1 | NP_001102572.1 | 1 | 343 | ||
| XM_008770771.1 | XP_008768993.1 | 3 | 1153 | |||
| XM_008770772.1 | XP_008768994.1 | 2 | 3228 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam167a| Gene Symbol: Fam167a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109102.1 | NP_001102572.1 | 1 | 343 | ||
| XM_008770771.1 | XP_008768993.1 | 3 | 1153 | |||
| XM_008770772.1 | XP_008768994.1 | 2 | 3228 | |||