siRNA Details
Gene Symbol: Akap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012773.2 | NP_036905.2 | 6 | 2232 | ||
XM_006252344.1 | XP_006252406.1 | 7 | 2406 | |||
XM_008770886.1 | XP_008769108.1 | 7 | 3298 | |||
XM_008770887.1 | XP_008769109.1 | 7 | 2488 | |||
XM_008770888.1 | XP_008769110.1 | 7 | 2488 | |||
XM_008770889.1 | XP_008769111.1 | 7 | 2488 | |||
XM_008770890.1 | XP_008769112.1 | 7 | 2488 |
