siRNA Details
| Gene Symbol: Commd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109105.1 | NP_001102575.1 | 3 | 391 | ||
| XM_008770891.1 | XP_008769113.1 | 4 | 305 | |||
| XM_008770892.1 | XP_008769114.1 | 2 | 298 | |||
| GenBank mRNA | FQ223060.1 | - | 3 | 191 | ||

Commd6
Genomic Map