siRNA Details
| Gene Symbol: Commd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109105.1 | NP_001102575.1 | 3, 4 | 425 | ||
| XM_008770891.1 | XP_008769113.1 | 4, 5 | 339 | |||
| XM_008770892.1 | XP_008769114.1 | 2, 3 | 332 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Commd6| Gene Symbol: Commd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109105.1 | NP_001102575.1 | 3, 4 | 425 | ||
| XM_008770891.1 | XP_008769113.1 | 4, 5 | 339 | |||
| XM_008770892.1 | XP_008769114.1 | 2, 3 | 332 | |||