siRNA Details
| Gene Symbol: Spert | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017506.1 | NP_001017506.1 | 3 | 1329 | ||
| XM_006252328.2 | XP_006252390.1 | 2 | 1599 | |||
| XM_006252329.2 | XP_006252391.1 | 3 | 1333 | |||
| XM_008770893.1 | XP_008769115.1 | 5 | 3769 | |||
| XM_008770894.1 | XP_008769116.1 | 4 | 1377 | |||
| XM_008770895.1 | XP_008769117.1 | 4 | 1370 | |||
| GenBank mRNA | BC079300.1 | - | 3 | 1329 | 7134526 | |

Spert
Genomic Map