siRNA Details
Gene Symbol: Ldb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110490.2 | NP_001103960.1 | 8, 9 | 854 | ||
NM_001277158.1 | NP_001264087.1 | 9, 10 | 869 | |||
NM_001277159.1 | NP_001264088.1 | 9, 10 | 859 | |||
XM_006252645.2 | XP_006252707.1 | 7, 8 | 1041 | |||
XM_006252647.1 | XP_006252709.1 | 8, 9 | 900 | |||
GenBank mRNA | BC083721.1 | - | 7, 8 | 792 | 7190925 | |
FM122871.1 | - | 6, 7 | 610 | |||
FQ214774.1 | - | 7, 8 | 699 | |||
FQ216341.1 | - | 8, 9 | 850 | |||
FQ216348.1 | - | 8, 9 | 849 | |||
FQ217970.1 | - | 9, 10 | 865 | |||
FQ223559.1 | - | 8, 9 | 848 |
