siRNA Details
| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128078.1 | NP_001121550.1 | 2 | 169 | ||
| GenBank mRNA | BC166765.1 | - | 2 | 169 | 7932317 | |
| FQ219981.1 | - | 2 | 191 | |||
| FQ229768.1 | - | 2 | 191 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam32a| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128078.1 | NP_001121550.1 | 2 | 169 | ||
| GenBank mRNA | BC166765.1 | - | 2 | 169 | 7932317 | |
| FQ219981.1 | - | 2 | 191 | |||
| FQ229768.1 | - | 2 | 191 | |||