siRNA Details
| Gene Symbol: Fam32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128078.1 | NP_001121550.1 | 2, 3 | 242 | ||
| GenBank mRNA | BC166765.1 | - | 2, 3 | 242 | 7932317 | |
| FQ219981.1 | - | 2, 3 | 264 | |||
| FQ229768.1 | - | 2, 3 | 264 | |||

Fam32a
Genomic Map