siRNA Details
| Gene Symbol: Fam129c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100908.1 | NP_001094378.1 | 13 | 1622 | ||
| GenBank mRNA | BC111077.1 | - | 13 | 1622 | ||
| FQ230626.1 | - | 5 | 585 | |||
| FQ231627.1 | - | 5 | 585 | |||
| FQ232480.1 | - | 5 | 585 | |||
| FQ233400.1 | - | 5 | 585 | |||

Fam129c
Genomic Map