siRNA Details
| Gene Symbol: Lrrc25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006252977.2 | XP_006253039.1 | 3 | 367 | ||
| XM_006252978.2 | XP_006253040.1 | 2 | 252 | |||
| XM_008771187.1 | XP_008769409.1 | 2 | 221 | |||
| XM_008771188.1 | XP_008769410.1 | 2 | 697 | |||
| XM_573882.5 | XP_573882.1 | 3 | 353 | |||
| GenBank mRNA | FQ228377.1 | - | 3 | 335 | ||

Lrrc25
Genomic Map