siRNA Details
| Gene Symbol: LOC364556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325145.1 | - | 382 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC364556| Gene Symbol: LOC364556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY325145.1 | - | 382 | |||