siRNA Details
| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109111.1 | NP_001102581.1 | 3 | 1126 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wwc2| Gene Symbol: Wwc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109111.1 | NP_001102581.1 | 3 | 1126 | ||