siRNA Details
| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191555.1 | NP_001178484.1 | 4 | 3132 | ||
| XM_008771331.1 | XP_008769553.1 | 4 | 3389 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip1| Gene Symbol: Rab11fip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191555.1 | NP_001178484.1 | 4 | 3132 | ||
| XM_008771331.1 | XP_008769553.1 | 4 | 3389 | |||