siRNA Details
| Gene Symbol: Adam5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020303.1 | NP_064699.1 | 8 | 697 | ||
| XM_006253321.2 | XP_006253383.1 | 8 | 747 | |||
| XM_006253322.2 | XP_006253384.1 | 8 | 684 | |||
| GenBank mRNA | BC079016.1 | - | 8 | 667 | ||
| BC091169.1 | - | 8 | 697 | 7112347 | ||
| Y11489.1 | - | 1 | 12 | |||

Adam5
Genomic Map