siRNA Details
| Gene Symbol: Tnfsf13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109112.1 | NP_001102582.1 | 2 | 723 | ||
| XM_006253480.2 | XP_006253542.1 | 2 | 1047 | |||
| XM_006253481.2 | XP_006253543.1 | 2 | 1047 | |||
| XM_006253482.2 | XP_006253544.1 | 2 | 1046 | |||
| XM_006253483.2 | XP_006253545.1 | 2 | 1045 | |||

Tnfsf13b
Genomic Map