siRNA Details
| Gene Symbol: MGC114246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017509.1 | NP_001017509.1 | 2 | 108 | ||
| GenBank mRNA | BC091563.1 | - | 2 | 108 | 7461240 | |
| Gene Symbol: Ctsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF456459.1 | - | 56 | |||

MGC114246
Genomic Map