siRNA Details
| Gene Symbol: Ctla2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109115.1 | NP_001102585.1 | 4 | 369 | ||
| XM_006253525.2 | XP_006253587.1 | 4 | 512 | |||
| GenBank mRNA | FQ220025.1 | - | 4 | 382 | ||
| FQ220118.1 | - | 4 | 382 | |||
| FQ220143.1 | - | 3 | 610 | |||
| FQ223122.1 | - | 2 | 149 | |||
| FQ223251.1 | - | 4 | 382 | |||
| FQ229997.1 | - | 4 | 313 | |||

Ctla2a
Genomic Map