siRNA Details
| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253530.1 | XP_006253592.1 | 4, 5 | 620 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctsll3| Gene Symbol: Ctsll3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006253530.1 | XP_006253592.1 | 4, 5 | 620 | ||