siRNA Details
| Gene Symbol: Cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126083.1 | NP_001119555.1 | 2, 3 | 303 | ||
| GenBank mRNA | BC162005.1 | - | 2, 3 | 303 | 9089459 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cks2| Gene Symbol: Cks2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126083.1 | NP_001119555.1 | 2, 3 | 303 | ||
| GenBank mRNA | BC162005.1 | - | 2, 3 | 303 | 9089459 | |