siRNA Details
| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109120.1 | NP_001102590.1 | 2 | 342 | ||
| GenBank mRNA | BC168934.1 | - | 2 | 299 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mboat1| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109120.1 | NP_001102590.1 | 2 | 342 | ||
| GenBank mRNA | BC168934.1 | - | 2 | 299 | ||