siRNA Details
| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109120.1 | NP_001102590.1 | 7 | 863 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mboat1| Gene Symbol: Mboat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109120.1 | NP_001102590.1 | 7 | 863 | ||