siRNA Details
| Gene Symbol: Tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034947.1 | NP_001030119.1 | 2 | 276 | ||
| GenBank mRNA | BC101920.1 | - | 2 | 276 | 7937874 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tdp2| Gene Symbol: Tdp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034947.1 | NP_001030119.1 | 2 | 276 | ||
| GenBank mRNA | BC101920.1 | - | 2 | 276 | 7937874 | |