siRNA Details
Gene Symbol: Tdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034947.1 | NP_001030119.1 | 3 | 400 | ||
GenBank mRNA | BC101920.1 | - | 3 | 400 | 7937874 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034947.1 | NP_001030119.1 | 3 | 400 | ||
GenBank mRNA | BC101920.1 | - | 3 | 400 | 7937874 |