siRNA Details
| Gene Symbol: LOC498750 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017510.3 | NP_001017510.1 | 1 | 536 | ||
| GenBank mRNA | AY325226.1 | - | 1 | 536 | ||
| BC085940.1 | - | 4 | 997 | 7123624 | ||
| BC089212.1 | - | 4 | 795 | 7302835 | ||
| Gene Symbol: LOC100911664 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751691.3 | XP_003751739.1 | 4 | 2712 | ||

LOC498750
Genomic Map