siRNA Details
| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109121.2 | NP_001102591.1 | 10 | 1091 | ||
| GenBank mRNA | BC168933.1 | - | 10 | 1091 | 7106331 | |
| FQ221736.1 | - | 10 | 1167 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stam| Gene Symbol: Stam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109121.2 | NP_001102591.1 | 10 | 1091 | ||
| GenBank mRNA | BC168933.1 | - | 10 | 1091 | 7106331 | |
| FQ221736.1 | - | 10 | 1167 | |||