siRNA Details
| Gene Symbol: Otud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574086.6 | XP_574086.3 | 1 | 2290 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Otud1| Gene Symbol: Otud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_574086.6 | XP_574086.3 | 1 | 2290 | ||