siRNA Details
| Gene Symbol: MGC116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024905.1 | NP_001020076.1 | 4 | 408 | ||
| GenBank mRNA | BC097984.1 | - | 4 | 408 | 7444421 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

MGC116121| Gene Symbol: MGC116121 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024905.1 | NP_001020076.1 | 4 | 408 | ||
| GenBank mRNA | BC097984.1 | - | 4 | 408 | 7444421 | |