siRNA Details
| Gene Symbol: Myo7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191941.1 | NP_001178870.1 | 22, 23 | 2929 | ||
| XM_006254600.2 | XP_006254662.1 | 22, 23 | 2985 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo7b| Gene Symbol: Myo7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191941.1 | NP_001178870.1 | 22, 23 | 2929 | ||
| XM_006254600.2 | XP_006254662.1 | 22, 23 | 2985 | |||