siRNA Details
Gene Symbol: Scgb3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024283.1 | NP_001019454.1 | 2 | 174 | ||
GenBank mRNA | AF439547.1 | - | 2 | 174 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scgb3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024283.1 | NP_001019454.1 | 2 | 174 | ||
GenBank mRNA | AF439547.1 | - | 2 | 174 |