siRNA Details
| Gene Symbol: Fam170a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126293.1 | NP_001119765.1 | 1 | 71 | ||
| XM_006254732.1 | XP_006254794.1 | 3 | 671 | |||
| XM_006254733.2 | XP_006254795.1 | 3 | 665 | |||
| XM_006254734.2 | XP_006254796.1 | 3 | 656 | |||
| XM_008772162.1 | XP_008770384.1 | 2 | 264 | |||
| GenBank mRNA | BC160906.1 | - | 3 | 657 | 7455403 | |

Fam170a
Genomic Map