siRNA Details
| Gene Symbol: RGD1562987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173472.1 | NP_001166943.1 | 2 | 157 | ||
| XM_008772163.1 | XP_008770385.1 | 2 | 135 | |||
| GenBank mRNA | BC162045.1 | - | 2 | 146 | 7107313 | |
| CB568734.1 | - | 2 | 158 | |||
| FQ217314.1 | - | 2 | 155 | |||
| FQ224078.1 | - | 2 | 145 | |||
| FQ228113.1 | - | 2 | 133 | |||
| FQ229472.1 | - | 2 | 115 | |||

RGD1562987
Genomic Map