siRNA Details
| Gene Symbol: RGD1562987 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173472.1 | NP_001166943.1 | 2 | 229 | ||
| XM_008772163.1 | XP_008770385.1 | 2 | 207 | |||
| GenBank mRNA | BC162045.1 | - | 2 | 218 | 7107313 | |
| CB568734.1 | - | 2 | 230 | |||
| FQ217314.1 | - | 2 | 227 | |||
| FQ224078.1 | - | 2 | 217 | |||
| FQ228113.1 | - | 2 | 205 | |||
| FQ229472.1 | - | 2 | 187 | |||

RGD1562987
Genomic Map