siRNA Details
| Gene Symbol: RGD1560212 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077676.1 | NP_001071144.1 | 4 | 952 | ||
| XM_006254957.2 | XP_006255019.1 | 3 | 907 | |||
| XM_008772164.1 | XP_008770386.1 | 4 | 928 | |||
| GenBank mRNA | BC127540.1 | - | 4 | 952 | 7301991 | |
| FQ234858.1 | - | 3 | 917 | |||

RGD1560212
Genomic Map