siRNA Details
| Gene Symbol: Dok6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191943.1 | NP_001178872.1 | 2, 3 | 214 | ||
| XM_008772165.1 | XP_008770387.1 | 1, 2 | 127 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dok6| Gene Symbol: Dok6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191943.1 | NP_001178872.1 | 2, 3 | 214 | ||
| XM_008772165.1 | XP_008770387.1 | 1, 2 | 127 | |||