siRNA Details
Gene Symbol: Dok6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191943.1 | NP_001178872.1 | 5 | 499 | ||
XM_008772165.1 | XP_008770387.1 | 4 | 412 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dok6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191943.1 | NP_001178872.1 | 5 | 499 | ||
XM_008772165.1 | XP_008770387.1 | 4 | 412 |