siRNA Details
| Gene Symbol: Arl2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024906.1 | NP_001020077.1 | 3 | 274 | ||
| XM_006255120.2 | XP_006255182.1 | 3 | 354 | |||
| XM_006255121.2 | XP_006255183.1 | 3 | 382 | |||
| GenBank mRNA | BC097448.1 | - | 3 | 274 | 7456585 | |

Arl2bp
Genomic Map